Assay Design



Gene Transcripts

Gene Symbol
MPV17
Entrez Gene ID
505763
Gene Name
mitochondrial inner membrane protein MPV17
Gene Aliases
-
Chromosome Location
Chr.11: 72324876 - 72334888 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045929.2 NP_001039394.1 3 - 4 249 63
GenBank mRNA BC112572.1 - 3 - 4 250 63

View Full Product Details