Assay Design



Gene Transcripts

Gene Symbol
PPP1R15A
Entrez Gene ID
514688
Gene Name
protein phosphatase 1 regulatory subunit 15A
Gene Aliases
-
Chromosome Location
Chr.18: 55474154 - 55477617 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046178.2 NP_001039643.1 2 - 3 1901 82
GenBank mRNA BC112768.1 - 2 - 3 1902 82

View Full Product Details