Assay Design



Gene Transcripts

Gene Symbol
NME8
Entrez Gene ID
518005
Gene Name
NME/NM23 family member 8
Gene Aliases
TXNDC3
Chromosome Location
Chr.4: 49680548 - 49730298 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046235.1 NP_001039700.1 4 - 5 296 105
GenBank mRNA BC110179.1 - 4 - 5 296 105

View Full Product Details