Assay Design



Gene Transcripts

Gene Symbol
HSD17B11
Entrez Gene ID
527592
Gene Name
hydroxysteroid 17-beta dehydrogenase 11
Gene Aliases
-
Chromosome Location
Chr.6: 102227494 - 102276972 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046286.2 NP_001039751.1 5 - 6 837 96
GenBank mRNA BC112527.1 - 5 - 6 838 96

View Full Product Details