Assay Design



Gene Transcripts

Gene Symbol
TRMT10C
Entrez Gene ID
532418
Gene Name
tRNA methyltransferase 10C, mitochondrial RNase P subunit
Gene Aliases
RG9MTD1
Chromosome Location
Chr.1: 45988248 - 45992514 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046323.2 NP_001039788.1 1 - 2 145 64
GenBank mRNA BC112774.1 - 1 - 2 147 64

View Full Product Details