Assay Design



Gene Transcripts

Gene Symbol
SUPT20H
Entrez Gene ID
534126
Gene Name
SPT20 homolog, SAGA complex component
Gene Aliases
FAM48A
Chromosome Location
Chr.12: 24646264 - 24685744 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046370.1 NP_001039835.1 4 - 5 237 115
GenBank mRNA BC105571.1 - 4 - 5 237 115

View Full Product Details