Assay Design



Gene Transcripts

Gene Symbol
FBXO28
Entrez Gene ID
537745
Gene Name
F-box protein 28
Gene Aliases
-
Chromosome Location
Chr.16: 27306978 - 27330899 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046425.1 NP_001039890.1 4 - 5 753 101
GenBank mRNA BC111213.1 - 4 - 5 753 101

View Full Product Details