Assay Design



Gene Transcripts

Gene Symbol
SCN3B
Entrez Gene ID
540925
Gene Name
sodium voltage-gated channel beta subunit 3
Gene Aliases
-
Chromosome Location
Chr.15: 34231050 - 34253814 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046495.1 NP_001039960.1 - 931 115
GenBank mRNA BC112810.1 - - 931 115

View Full Product Details