Assay Design



Gene Transcripts

Gene Symbol
LY96
Entrez Gene ID
613753
Gene Name
lymphocyte antigen 96
Gene Aliases
MD-2, MD2, ly-96
Chromosome Location
Chr.14: 37241063 - 37273887 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046517.1 NP_001039982.1 2 - 3 207 104
GenBank mRNA AB072456.1 - 2 - 3 223 104
DQ319076.1 - 2 - 3 207 104

View Full Product Details