Assay Design



Gene Transcripts

Gene Symbol
BNIP1
Entrez Gene ID
614520
Gene Name
BCL2 interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.20: 4841496 - 4859976 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046539.5 NP_001040004.2 2 - 3 189 152
GenBank mRNA BC110132.1 - 2 - 3 193 152
EE229689.1 - 2 - 3 190 152

View Full Product Details