Assay Design



Gene Transcripts

Gene Symbol
CXCL10
Entrez Gene ID
615107
Gene Name
C-X-C motif chemokine ligand 10
Gene Aliases
-
Chromosome Location
Chr.6: 90881993 - 90884345 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046551.2 NP_001040016.1 1 - 2 121 68
GenBank mRNA BC112547.1 - 1 - 2 122 68
EU276062.1 - 1 - 2 69 68

View Full Product Details