Assay Design



Gene Transcripts

Gene Symbol
COX16
Entrez Gene ID
615979
Gene Name
cytochrome c oxidase assembly factor COX16
Gene Aliases
C10H14ORF112
Chromosome Location
Chr.10: 82020602 - 82060892 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046568.1 NP_001040033.1 3 - 4 325 151
GenBank mRNA BC111674.1 - 3 - 4 325 151

View Full Product Details