Assay Design



Gene Transcripts

Gene Symbol
CCK
Entrez Gene ID
617510
Gene Name
cholecystokinin
Gene Aliases
-
Chromosome Location
Chr.22: 14568045 - 14573729 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046603.2 NP_001040068.1 2 - 3 272 67
GenBank mRNA BC114184.1 - 2 - 3 273 67

View Full Product Details