Assay Design



Gene Transcripts

Gene Symbol
ATP6V1D
Entrez Gene ID
404152
Gene Name
ATPase H+ transporting V1 subunit D
Gene Aliases
VATD
Chromosome Location
Chr.10: 79239415 - 79255202 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075141.2 NP_001068609.2 5 - 6 493 86
NM_001257088.1 NP_001244017.1 4 - 5 375 86
GenBank mRNA AB098994.1 - 5 - 6 431 86
BC105495.1 - 5 - 6 440 86
EH151848.1 - 5 - 6 494 86
FE012554.1 - 4 - 5 282 86

View Full Product Details