Assay Design



Gene Transcripts

Gene Symbol
ZFPL1
Entrez Gene ID
613442
Gene Name
zinc finger protein like 1
Gene Aliases
-
Chromosome Location
Chr.29: 43297356 - 43301446 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076272.1 NP_001069740.1 3 - 4 379 142
GenBank mRNA BC110275.1 - 3 - 4 379 142
BT025470.1 - 3 - 4 409 142

View Full Product Details