Assay Design



Gene Transcripts

Gene Symbol
YPEL2
Entrez Gene ID
616180
Gene Name
yippee like 2
Gene Aliases
-
Chromosome Location
Chr.19: 10299237 - 10366627 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079510.2 NP_001072978.1 2 - 3 454 64
GenBank mRNA BC110210.1 - 2 - 3 455 64

View Full Product Details