Assay Design



Gene Transcripts

Gene Symbol
LIN7A
Entrez Gene ID
528379
Gene Name
lin-7 homolog A, crumbs cell polarity complex component
Gene Aliases
-
Chromosome Location
Chr.5: 10346168 - 10506559 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079602.2 NP_001073070.1 3 - 4 484 77
GenBank mRNA BC109509.1 - 3 - 4 485 77

View Full Product Details