Assay Design



Gene Transcripts

Gene Symbol
MED30
Entrez Gene ID
613879
Gene Name
mediator complex subunit 30
Gene Aliases
THRAP6, Trap25
Chromosome Location
Chr.14: 46782520 - 46801620 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079618.2 NP_001073086.1 4 - 5 695 66
GenBank mRNA BC110250.1 - 4 - 5 696 66

View Full Product Details