Assay Design



Gene Transcripts

Gene Symbol
ITGB3BP
Entrez Gene ID
614469
Gene Name
integrin subunit beta 3 binding protein
Gene Aliases
-
Chromosome Location
Chr.3: 82052235 - 82157908 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079619.2 NP_001073087.1 7 - 8 565 162
GenBank mRNA BC112592.1 - 7 - 8 566 162

View Full Product Details