Assay Design
Gene Transcripts
- Gene Symbol
- C4BPB
- Entrez Gene ID
-
281652
- Gene Name
- complement component 4 binding protein beta
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 4913201 - 4925089 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_174253.3 |
NP_776678.2 |
4 - 5 |
728 |
157 |
GenBank mRNA |
BC112543.1 |
- |
4 - 5 |
731 |
157 |
View Full Product Details