Assay Design



Gene Transcripts

Gene Symbol
PBLD
Entrez Gene ID
504417
Gene Name
phenazine biosynthesis like protein domain containing
Gene Aliases
-
Chromosome Location
Chr.28: 24716173 - 24751226 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045886.2 NP_001039351.1 3 - 4 315 94
GenBank mRNA BC105465.1 - 3 - 4 316 94

View Full Product Details