Assay Design



Gene Transcripts

Gene Symbol
NAB2
Entrez Gene ID
504755
Gene Name
NGFI-A binding protein 2
Gene Aliases
-
Chromosome Location
Chr.5: 56340387 - 56346415 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045897.1 NP_001039362.1 6 - 7 1630 71
GenBank mRNA BC114714.1 - 6 - 7 1630 71

View Full Product Details