Assay Design



Gene Transcripts

Gene Symbol
DPT
Entrez Gene ID
504963
Gene Name
dermatopontin
Gene Aliases
-
Chromosome Location
Chr.16: 36297417 - 36333818 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045903.1 NP_001039368.1 1 - 2 301 94
GenBank mRNA BC113221.1 - 1 - 2 301 94

View Full Product Details