Assay Design



Gene Transcripts

Gene Symbol
NUDT13
Entrez Gene ID
504993
Gene Name
nudix hydrolase 13
Gene Aliases
-
Chromosome Location
Chr.28: 29189839 - 29208002 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045905.2 NP_001039370.1 1 - 2 88 82
GenBank mRNA BC114867.1 - 1 - 2 89 82

View Full Product Details