Assay Design



Gene Transcripts

Gene Symbol
MRPL53
Entrez Gene ID
505728
Gene Name
mitochondrial ribosomal protein L53
Gene Aliases
-
Chromosome Location
Chr.11: 10159213 - 10162300 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045926.2 NP_001039391.1 1 - 2 112 92
GenBank mRNA BC105487.1 - 1 - 2 113 92

View Full Product Details