Assay Design



Gene Transcripts

Gene Symbol
NCDN
Entrez Gene ID
505994
Gene Name
neurochondrin
Gene Aliases
-
Chromosome Location
Chr.3: 110202070 - 110210968 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045931.1 NP_001039396.1 1 - 2 89 91
GenBank mRNA BC105450.1 - 1 - 2 89 91

View Full Product Details