Assay Design



Gene Transcripts

Gene Symbol
CMPK1
Entrez Gene ID
509965
Gene Name
cytidine/uridine monophosphate kinase 1
Gene Aliases
CK, CMPK
Chromosome Location
Chr.3: 98887957 - 98924195 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046044.1 NP_001039509.1 2 - 3 333 66
GenBank mRNA BC112478.1 - 2 - 3 333 66

View Full Product Details