Assay Design



Gene Transcripts

Gene Symbol
HIBADH
Entrez Gene ID
512002
Gene Name
3-hydroxyisobutyrate dehydrogenase
Gene Aliases
-
Chromosome Location
Chr.4: 68473893 - 68582089 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046106.1 NP_001039571.1 3 - 4 365 121
GenBank mRNA BC105543.1 - 3 - 4 365 121

View Full Product Details