Assay Design



Gene Transcripts

Gene Symbol
TVP23B
Entrez Gene ID
513550
Gene Name
trans-golgi network vesicle protein 23 homolog B
Gene Aliases
FAM18B, FAM18B1, FAM18B2
Chromosome Location
Chr.19: 32983940 - 32993498 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046152.2 NP_001039617.1 2 - 3 205 130
GenBank mRNA BC113231.1 - 2 - 3 101 130
EH153594.1 - 2 - 3 205 130

View Full Product Details