Assay Design



Gene Transcripts

Gene Symbol
C14H8orf33
Entrez Gene ID
513585
Gene Name
chromosome 14 C8orf33 homolog
Gene Aliases
C8orf33
Chromosome Location
Chr.14: 303633 - 305704 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046153.3 NP_001039618.2 1 - 2 52 69
GenBank mRNA BC112675.1 - 1 - 2 36 69
EH210109.1 - 1 - 2 53 69

View Full Product Details