Assay Design



Gene Transcripts

Gene Symbol
MDM4
Entrez Gene ID
514225
Gene Name
MDM4 regulator of p53
Gene Aliases
-
Chromosome Location
Chr.16: 2253349 - 2305299 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046169.1 NP_001039634.1 8 - 9 753 78
GenBank mRNA BC113350.1 - 8 - 9 753 78

View Full Product Details