Assay Design



Gene Transcripts

Gene Symbol
TBC1D24
Entrez Gene ID
529002
Gene Name
TBC1 domain family member 24
Gene Aliases
-
Chromosome Location
Chr.25: 1979510 - 2006263 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046296.2 NP_001039761.1 2 - 3 149 66
GenBank mRNA BC114148.1 - 2 - 3 150 66

View Full Product Details