Assay Design



Gene Transcripts

Gene Symbol
THOC3
Entrez Gene ID
529231
Gene Name
THO complex 3
Gene Aliases
-
Chromosome Location
Chr.10: 5186072 - 5197306 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046300.2 NP_001039765.1 4 - 5 854 69
GenBank mRNA BC114172.1 - 4 - 5 855 69

View Full Product Details