Assay Design



Gene Transcripts

Gene Symbol
BAMBI
Entrez Gene ID
530147
Gene Name
BMP and activin membrane bound inhibitor
Gene Aliases
-
Chromosome Location
Chr.13: 36021690 - 36028577 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046309.1 NP_001039774.1 1 - 2 381 76
GenBank mRNA BC114655.1 - 1 - 2 381 76

View Full Product Details