Assay Design



Gene Transcripts

Gene Symbol
MPHOSPH10
Entrez Gene ID
531149
Gene Name
M-phase phosphoprotein 10
Gene Aliases
-
Chromosome Location
Chr.21: 27396948 - 27409259 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046317.2 NP_001039782.1 1 - 2 122 120
GenBank mRNA BC112826.1 - 1 - 2 123 120

View Full Product Details