Assay Design



Gene Transcripts

Gene Symbol
SLC10A1
Entrez Gene ID
532890
Gene Name
solute carrier family 10 member 1
Gene Aliases
-
Chromosome Location
Chr.10: 81503835 - 81526722 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046339.1 NP_001039804.1 4 - 5 1051 91
GenBank mRNA BC105471.1 - 4 - 5 1051 91
BC146029.1 - 4 - 5 1254 91

View Full Product Details