Assay Design



Gene Transcripts

Gene Symbol
PQBP1
Entrez Gene ID
534894
Gene Name
polyglutamine binding protein 1
Gene Aliases
-
Chromosome Location
Chr.X: 87013764 - 87019563 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046389.1 NP_001039854.1 1 - 2 612 76
GenBank mRNA BC105573.1 - 1 - 2 612 76

View Full Product Details