Assay Design



Gene Transcripts

Gene Symbol
PPM1K
Entrez Gene ID
540329
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1K
Gene Aliases
-
Chromosome Location
Chr.6: 36449102 - 36472317 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046474.2 NP_001039939.1 1 - 2 154 89
GenBank mRNA AJ871967.1 - 1 - 2 128 89
AJ871968.1 - 1 - 2 128 89
BC118079.1 - 1 - 2 155 89

View Full Product Details