Assay Design



Gene Transcripts

Gene Symbol
PPM1K
Entrez Gene ID
540329
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1K
Gene Aliases
-
Chromosome Location
Chr.6: 36449102 - 36472317 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046474.2 NP_001039939.1 6 - 7 1215 82
GenBank mRNA AJ871967.1 - 6 - 7 1189 82
AJ871968.1 - 6 - 7 1045 82
BC118079.1 - 6 - 7 1216 82

View Full Product Details