Assay Design



Gene Transcripts

Gene Symbol
IFT46
Entrez Gene ID
505579
Gene Name
intraflagellar transport 46
Gene Aliases
C11orf60, C15H11orf60
Chromosome Location
Chr.15: 29204163 - 29219665 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075209.1 NP_001068677.1 1 - 2 99 103
GenBank mRNA BC116107.1 - 1 - 2 99 103

View Full Product Details