Assay Design



Gene Transcripts

Gene Symbol
MBLAC1
Entrez Gene ID
505636
Gene Name
metallo-beta-lactamase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.25: 36331651 - 36333031 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075212.1 NP_001068680.1 1 - 1 1252 65
GenBank mRNA BC113225.1 - 1 - 1 1252 65

View Full Product Details