Assay Design



Gene Transcripts

Gene Symbol
BCDIN3D
Entrez Gene ID
505650
Gene Name
BCDIN3 domain containing RNA methyltransferase
Gene Aliases
-
Chromosome Location
Chr.5: 30042235 - 30047718 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075216.1 NP_001068684.1 1 - 2 231 94
GenBank mRNA BC113219.1 - 1 - 2 231 94

View Full Product Details