Assay Design



Gene Transcripts

Gene Symbol
RDH13
Entrez Gene ID
508082
Gene Name
retinol dehydrogenase 13
Gene Aliases
-
Chromosome Location
Chr.18: 62338040 - 62355023 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075345.2 NP_001068813.1 6 - 7 878 72
GenBank mRNA AM930554.2 - 4 - 5 459 72
BC118441.1 - 6 - 7 879 72

View Full Product Details