Assay Design



Gene Transcripts

Gene Symbol
SNCB
Entrez Gene ID
508421
Gene Name
synuclein beta
Gene Aliases
-
Chromosome Location
Chr.7: 38091556 - 38100473 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075360.1 NP_001068828.1 1 - 2 243 156
GenBank mRNA BC118378.1 - 1 - 2 243 156

View Full Product Details