Assay Design



Gene Transcripts

Gene Symbol
DBX2
Entrez Gene ID
510669
Gene Name
developing brain homeobox 2
Gene Aliases
-
Chromosome Location
Chr.5: 35292206 - 35329400 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075461.1 NP_001068929.1 2 - 3 811 101
GenBank mRNA BC118220.1 - 2 - 3 811 101

View Full Product Details