Assay Design



Gene Transcripts

Gene Symbol
CA1
Entrez Gene ID
510801
Gene Name
carbonic anhydrase I
Gene Aliases
-
Chromosome Location
Chr.14: 77144212 - 77154651 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075466.1 NP_001068934.1 6 - 7 743 89
GenBank mRNA BC116126.1 - 6 - 7 743 89

View Full Product Details