Assay Design



Gene Transcripts

Gene Symbol
PIK3IP1
Entrez Gene ID
512082
Gene Name
phosphoinositide-3-kinase interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.17: 70094432 - 70105120 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075552.1 NP_001069020.1 1 - 2 171 59
GenBank mRNA BC114715.1 - 1 - 2 171 59
BT025449.1 - 1 - 2 188 59

View Full Product Details