Assay Design



Gene Transcripts

Gene Symbol
NUDT16
Entrez Gene ID
512320
Gene Name
nudix hydrolase 16
Gene Aliases
-
Chromosome Location
Chr.1: 138548856 - 138550443 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075560.1 NP_001069028.1 1 - 2 135 64
GenBank mRNA BC126814.1 - 1 - 2 158 64
BT025346.1 - 1 - 2 135 64

View Full Product Details