Assay Design



Gene Transcripts

Gene Symbol
CLBA1
Entrez Gene ID
512628
Gene Name
clathrin binding box of aftiphilin containing 1
Gene Aliases
C14orf79, C21H14orf79
Chromosome Location
Chr.21: 69403996 - 69409670 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075573.1 NP_001069041.1 4 - 5 1032 85
GenBank mRNA BC118243.1 - 4 - 5 1032 85

View Full Product Details