Assay Design



Gene Transcripts

Gene Symbol
KCNIP3
Entrez Gene ID
513316
Gene Name
potassium voltage-gated channel interacting protein 3
Gene Aliases
KChIP3
Chromosome Location
Chr.11: 2042314 - 2136545 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075612.2 NP_001069080.1 4 - 5 453 86
GenBank mRNA BC118417.1 - 4 - 5 454 86

View Full Product Details